Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01629

Millipore

Terrific broth

suitable for microbiology

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$128.00
5 KG
$1,210.00

$128.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
500 G
$128.00
5 KG
$1,210.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

$128.00


Please contact Customer Service for Availability

Request more information

sterility

non-sterile

Quality Level

form

granular

manufacturer/tradename

GranuCult® plus

technique(s)

microbiological culture: suitable

application(s)

clinical
pharmaceutical

microbiology

storage temp.

15-25°C

suitability

Escherichia coli

General description

for microbiology

Linkage

Replaces: 1016295007

Analysis Note

Appearance (clearness): clear
Appearance (colour): yellowish-brown
pH-value (25 °C): 7.0 - 7.4
Colony count (Escherichia coli ATCC 23724): ≥ 1E+08 CFU/ml
Colony count (Escherichia coli ATCC 33694): ≥ 1E+08 CFU/ml
Colony count (Escherichia coli ATCC 39403 (JM103)): ≥ 1E+08 CFU/ml
Colony count (Escherichia coli ATCC 47014): ≥ 1E+08 CFU/ml
Colony count (Escherichia coli ATCC 47013): ≥ 1E+08 CFU/ml
Colony count (Escherichia coli ATCC 53868 (DH5a)): ≥ 1E+08 CFU/ml
Incubation: 24 hrs.; 35 °C; aerobically.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service