Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01374

Sigma-Aldrich

Brilliant green (hydrogen sulfate) (C.I. 42040)

for microscopy Certistain®

Synonym(s):

Brilliant green (hydrogen sulfate) (C.I. 42040), Diamond green G, Solid green, Ethyl green

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H34N2O4S
CAS Number:
Molecular Weight:
482.63
MDL number:
UNSPSC Code:
41116130
EC Index Number:
211-190-1
Pricing and availability is not currently available.

Quality Level

form

solid

potency

300-400 mg/kg LD50, oral (Rat)

composition

Dye, ≥95% spectrophotometric assay

pH

2.5 (25 °C, 10 g/L in H2O)

mp

210 °C (decomposition)

solubility

100 g/L

bulk density

500‑600 kg/m3

εmax

2210-2350 at 628-632 nm in 50% ethanol at 0.003 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C27H33N2.H2O4S/c1-5-28(6-2)25-18-14-23(15-19-25)27(22-12-10-9-11-13-22)24-16-20-26(21-17-24)29(7-3)8-4;1-5(2,3)4/h9-21H,5-8H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

NNBFNNNWANBMTI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Brilliant green (hydrogen sulfate) (C.I. 42040) for microscopy Certistain®, is a dry staining dye that is used to prepare a staining solution makes target structures in bacteriological specimen materials evaluable for diagnostic purposes. In the area of bacteriology, among others the Gram staining method, the Ziehl-Neelsen staining method, and the spore-staining method according to Rakette are used to yield a differentiated image of pathogens. This can be done either by using eosin Y as a counterstain or by modification according to Wirtz using safranine O solution. By this method, spores are always stained green and the cell elements always red.
As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Dye content (spectrophotometrically): ≥ 95 %
Identity
- Hydrogen sulfate (as sulfur):
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (ethanol 50 %): 628 - 632 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.003 g/l; ethanol 50 %): 2210 - 2350
TLC-Test: passes test
Loss on drying (110 °C): ≤ 5 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service