Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01231

Supelco

Isoamyl acetate

EMPLURA®

Synonym(s):

Isoamyl acetate, iso-Amyl acetate, Acetic acid isoamyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O2
CAS Number:
Molecular Weight:
130.18
MDL number:
UNSPSC Code:
12191502
EC Index Number:
204-662-3

vapor pressure

7 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥99% (GC)

form

liquid

autoignition temp.

355 °C

potency

>5000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rat)

expl. lim.

1-10 % (v/v)

impurities

≤0.1% Free acid (as CH3COOH)
≤0.1% Water
≤1% Isoamyl alcohol (GC)

pH

7

bp

142 °C/1013 hPa

mp

-78 °C

transition temp

flash point 36 °C

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H14O2/c1-6(2)4-5-9-7(3)8/h6H,4-5H2,1-3H3

InChI key

MLFHJEHSLIIPHL-UHFFFAOYSA-N

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Free acid (as CH₃COOH): ≤ 0.1 %
Density (d 20 °C/ 4 °C): 0.872 - 0.873
Isoamyl alcohol (GC): ≤ 1.0 %
Water: ≤ 0.1 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service