Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01118

Supelco

Ammonium benzoate

EMPLURA®

Synonym(s):

Ammonium benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NO2
CAS Number:
Molecular Weight:
139.15
MDL number:
UNSPSC Code:
12352107
EC Index Number:
217-468-9
Assay:
≥98.0% (perchloric acid titration)
form:
solid
solubility:
200 g/L

Quality Level

product line

EMPLURA®

Assay

≥98.0% (perchloric acid titration)

form

solid

potency

825 mg/kg LD50, oral (Rat)

pH

6.4 (20 °C, 100 g/L in H2O)

mp

198 °C (decomposition)

solubility

200 g/L

density

1.26 g/cm3 at 25 °C

cation traces

heavy metals (as Pb): ≤0.001%

storage temp.

2-30°C

InChI

1S/C7H6O2.H3N/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);1H3

InChI key

VWSRWGFGAAKTQG-UHFFFAOYSA-N

Analysis Note

Assay (perchloric acid titration): ≥ 98.0 %
Identity (IR-spectrum): passes test
pH-value (10 %; water): 6.0 - 8.0
Heavy metals (as Pb): ≤ 0.001 %
Oxidizable matter: passes test
Residual solvents (Ph Eur/ICH): excluded by production process

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service