Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01063

Supelco

Aluminium nitrate nonahydrate

for analysis EMSURE®

Synonym(s):

Aluminium nitrate nonahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$249.00
50 KG
$12,610.00

$249.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 G
$249.00
50 KG
$12,610.00

About This Item

Linear Formula:
Al(NO3)3 · 9 H2O
CAS Number:
Molecular Weight:
375.13
MDL number:
UNSPSC Code:
12352302
EC Index Number:
236-751-8
NACRES:
NA.21
form:
solid
solubility:
41.9 g/L

$249.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

EMSURE®

Assay

≥98.5% (complexometric)

form

solid

potency

3671 mg/kg LD50, oral (Rat)

pH

2.0-4.0 (20 °C, 50 g/L in H2O)

mp

73 °C

solubility

41.9 g/L

density

1.72 g/cm3 at 20 °C

bulk density

880 kg/m3

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.002%
K: ≤0.002%
NH4+: ≤0.02%
Na: ≤0.005%
heavy metals (as Pb): ≤0.001%

storage temp.

2-30°C

SMILES string

[Al]1(O[N+](=O)O1)O[N+](=O)[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O

InChI

1S/Al.3NO3.9H2O/c;3*2-1(3)4;;;;;;;;;/h;;;;9*1H2/q+3;3*-1;;;;;;;;;

InChI key

SWCIQHXIXUMHKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • A thiazole-based colorimetric and photoluminescent chemosensors for As(3+) ions detection: This application highlights the use of arsenic tests in developing sensitive chemosensors that are crucial for monitoring arsenic levels in various environments, ensuring public health and safety (Dhanasekaran K et al., 2024).



Analysis Note

Assay (complexometric): ≥ 98.5 %
pH-value (5 %; water): 2.0 - 4.0
Chloride (Cl): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.002 %
K (Potassium): ≤ 0.002 %
Na (Sodium): ≤ 0.005 %
NH₄ (Ammonium): ≤ 0.02 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service