Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.01036

Supelco

Chromium(III) potassium sulfate dodecahydrate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Chromium(III) potassium sulfate dodecahydrate, Potassium chromium(III) sulfate, Alum chrome

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCr(SO4)2 · 12H2O
CAS Number:
Molecular Weight:
499.40
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-401-6
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

99.0-102.0% (iodometric)

form

solid

impurities

≤0.01% Insoluble matter

pH

3 (20 °C, 50 g/L in H2O)

mp

89 °C

density

1.83 g/cm3 at 20 °C

bulk density

800‑1000 kg/m3

anion traces

chloride (Cl-): ≤0.002%

cation traces

Al: ≤0.005%
Cu: ≤0.001%
Fe: ≤0.010%
NH4+: ≤0.010%
Pb: ≤0.005%
heavy metals (as Pb): ≤0.01%

storage temp.

15-25°C

InChI

1S/Cr.K.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;;2*(H2,1,2,3,4);12*1H2/q+3;+1;;;;;;;;;;;;;;/p-4

InChI key

ZFVHBEKVAITXHW-UHFFFAOYSA-J

Analysis Note

Assay (iodometric): 99.0 - 102.0 %
Identity: passes test
Insoluble matter: ≤ 0.01 %
Chloride (Cl): ≤ 0.002 %
Heavy metals (as Pb): ≤ 0.01 %
Al (Aluminium): ≤ 0.005 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.010 %
NH₄ (Ammonium): ≤ 0.010 %
Pb (Lead): ≤ 0.005 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service