Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01008

Millipore

ß-Alanine

for biochemistry

Synonym(s):

ß-Alanine, Ala, 3-Aminopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7NO2
CAS Number:
Molecular Weight:
89.09
MDL number:
UNSPSC Code:
12352209
EC Index Number:
203-536-5
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

form

solid

pH

6.5-7.5 (20 °C, 100 g/L in H2O)

mp

200 °C (decomposition)

density

1.44 g/cm3 at 20 °C

bulk density

770 kg/m3

storage temp.

2-30°C

SMILES string

NCCC(=O)O

InChI

1S/C3H7NO2/c4-2-1-3(5)6/h1-2,4H2,(H,5,6)

InChI key

UCMIRNVEIXFBKS-UHFFFAOYSA-N

Application


  • Non-proteinogenic amino acids mitigate oxidative stress and enhance the resistance of common bean plants against Sclerotinia sclerotiorum.: Demonstrating the utility of ß-Alanine in plant science, this study explores its role in enhancing plant resistance to pathogens by modulating oxidative stress responses. The findings contribute to the development of more resilient agricultural practices (Nehela Y et al., 2024).

Analysis Note

Assay (perchloric acid titration): ≥ 99
Identity (IR-spectrum): passes test
Heavy metals (as Pb): ≤ 0.001 %
TLC-Test: passes test

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service