Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.00886

SAFC

Ethyl 4-hydroxybenzoate sodium salt

EMPROVE® ESSENTIAL, Ph. Eur., BP, NF

Pharma Manufacturing

Synonym(s):

Ethyl 4-hydroxybenzoate sodium salt, p-Hydroxybenzoic acid ethyl ester sodium salt, Ethylparaben sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 KG
$1,206.00

$1,206.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 KG
$1,206.00

About This Item

Empirical Formula (Hill Notation):
C9H9NaO3
CAS Number:
Molecular Weight:
188.16
MDL number:
UNSPSC Code:
12352108
EC Index Number:
252-487-6

$1,206.00


Please contact Customer Service for Availability

Request a Bulk Order

Agency

BP
NF
Ph. Eur.

Quality Level

product line

EMPROVE® ESSENTIAL

form

solid

pH

10 (20 °C, 1 g/L in H2O)

bp

297 °C/1013 hPa (free ester,decomposition)

mp

>115 °C (free ester)

transition temp

flash point 121 °C

solubility

1000 g/L

application(s)

liquid formulation
ophthalmics
pharmaceutical

storage temp.

15-25°C

SMILES string

[Na+].[O-]c1ccc(cc1)C(=O)OCC

InChI

1S/C9H10O3.Na/c1-2-12-9(11)7-3-5-8(10)6-4-7;/h3-6,10H,2H2,1H3;/q;+1/p-1

InChI key

QYNMSPKSYXPZHG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Ethyl 4-hydroxybenzoate has antimicrobial characteristics and is used as preservative in pharmaceutical formulations and cosmetics. It can be combined with other parabens like methyl 4-hydroxybenzoate or propyl 4-hydroxybenzoate. The sodium salt provides improved solubility in water compared to standard ethyl 4-hydroxybenzoate.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

249.8 °F

Flash Point(C)

121 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service