Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.00803

SAFC

L(+)-Tartaric acid

powder, EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JP, NF, E 334

Pharma Manufacturing

Synonym(s):

L(+)-Tartaric acid, 2,3-Dihydroxybutanedioic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 KG
$1,918.00

$1,918.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest a Sample

Select a Size

Change View
25 KG
$1,918.00

About This Item

Linear Formula:
HOOCCH(OH)CH(OH)COOH
CAS Number:
Molecular Weight:
150.09
MDL number:
UNSPSC Code:
12352106
E Number:
E334
EC Index Number:
201-766-0

$1,918.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest a Sample

Agency

BP
ChP
JP
NF
Ph. Eur.

Quality Level

vapor pressure

<5 Pa ( 20 °C)

product line

EMPROVE® ESSENTIAL

Assay

99.7-100.5% dry basis (acidimetric)

form

powder

autoignition temp.

425 °C

technique(s)

API processing | cocrystal formation: suitable
API processing | salt formation: suitable

pH

1.6 (25 °C, 100 g/L in H2O)

mp

168-170 °C

transition temp

flash point 210 °C

solubility

1390 g/L

density

1.76 g/cm3 at 20 °C

bulk density

1000 kg/m3

cation traces

As: ≤0.0001%
Ca: ≤0.0200%
Ca:, passes test
Hg: ≤0.0001%
Pb: ≤0.0002%
heavy metals (as Pb): ≤0.0010%

application(s)

liquid formulation
pharmaceutical
semi-solid formulation
solid formulation

storage temp.

2-30°C

SMILES string

[O-]C(=O)C(O)C(O)C(=O)[O-].[H+].[H+]

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)

InChI key

FEWJPZIEWOKRBE-UHFFFAOYSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Tartaric acid is a commonly used ingredient in both food and pharmaceutics. In pharmaceutical applications, tartaric acid has been used primarily as a pH modifier, and in the production of effervescent formulations.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service