Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.00659

Supelco

Tungstosilicic acid hydrate

for analysis EMSURE®

Synonym(s):

Tungstosilicic acid hydrate, Silicotungstic acid, Wolframosilicic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H4[Si(W3O10)4] · x H2O
CAS Number:
Molecular Weight:
2878.17 (anhydrous basis)
MDL number:
UNSPSC Code:
12352301
EC Index Number:
234-719-8
NACRES:
NA.21

Quality Level

product line

EMSURE®

form

solid

impurities

≤20 ppm Total nitrogen (N)

loss

≤16% loss on ignition, 750 °C

solubility

water: soluble at 20 °C

bulk density

200‑500 kg/m3

anion traces

chloride (Cl-): ≤20 ppm
sulfate (SO42-): ≤50 ppm

cation traces

Fe: ≤10 ppm
K: ≤200 ppm
Na: ≤200 ppm
heavy metals (as Pb): ≤20 ppm

storage temp.

15-25°C

InChI

1S/H4O4Si.H2O.36O.12W/c1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h1-4H;1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;

InChI key

ACTPFSYIGLFGLY-UHFFFAOYSA-N

Analysis Note

Identity: passes test
Chloride (Cl): ≤ 20 ppm
Sulphate (SO₄): ≤ 50 ppm
Total nitrogen (N): ≤ 20 ppm
Heavy metals (as Pb): ≤ 20 ppm
Fe (Iron): ≤ 10 ppm
K (Potassium): ≤ 200 ppm
Na (Sodium): ≤ 200 ppm
Loss on ignition (750 °C): ≤ 16 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service