Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.00640

Millipore

Acrylamide-bis ready-to-use solution 40% (19:1)

for electrophoresis

Synonym(s):

40% Acrylamide/Bisacrylamide 19:1, Polyacrylamide solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105323

Quality Level

density

1.03 g/cm3

storage temp.

2-8°C

Analysis Note

Appearance of solution (color): almost colourless
Appearance of solution (clearness): almost clear
Assay of N,N′-methylene-bisacrylamide (HPLC): 4.8 - 5.2 %
Spec. Absorptivity A 1%/1cm (290 nm; 1 %; 1 cm; water): ≤ 0.15
Free acid (as acrylic acid): ≤ 0.002 %

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service