Skip to Content
MilliporeSigma
All Photos(2)

Documents

1.00514

Supelco

Perchloric acid

70%, for analysis (max. 0.0000005% Hg) EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Perchloric acid, PCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HClO4
CAS Number:
Molecular Weight:
100.46
MDL number:
UNSPSC Code:
12352301
NACRES:
NA.21

product name

Perchloric acid 70%, for analysis (max. 0.0000005% Hg) EMSURE® ACS,ISO,Reag. Ph Eur

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

product line

EMSURE®

form

liquid

concentration

70.0-72.0% HClO4

impurities

≤0.005 ppm Hg (Mercury)
≤0.02 ppm Ba (Barium)
≤0.02 ppm Be (Beryllium)
≤0.02 ppm Li (Lithium)
≤0.02 ppm Mn (Manganese)
≤0.02 ppm Sr (Strontium)
≤0.05 ppm Al (Aluminium)
≤0.05 ppm As (Arsenic)
≤0.05 ppm Cd (Cadmium)
≤0.05 ppm Co (Cobalt)
≤0.05 ppm Ge (Germanium)
≤0.05 ppm Mo (Molybdenum)
≤0.05 ppm Pb (Lead)
≤0.05 ppm Tl (Thallium)
≤0.05 ppm V (Vanadium)
≤0.1 ppm Ag (Silver)
≤0.1 ppm Bi (Bismuth)
≤0.1 ppm Cu (Copper)
≤0.1 ppm K (Potassium)
≤0.1 ppm Ni (Nickel)
≤0.1 ppm Ti (Titanium)
≤0.1 ppm Zn (Zinc)
≤0.1 ppm Zr (Zirconium)
≤0.5 ppm Ca (Calcium)
≤0.5 ppm Free chlorine
≤0.5 ppm Mg (Magnesium)
≤0.5 ppm Na (Sodium)
≤1 ppm Heavy metals (as Pb)
≤1.0 ppm Fe (Iron)
≤10 ppm Chlorate (ClO3)
≤10 ppm Sulfate (SO4)
≤10 ppm Total nitrogen (N)
≤3 ppm Chloride (Cl)
≤5 ppm Phosphate and Silicate (as SiO2)

ign. residue

≤30 ppm (as sulfate)

SMILES string

OCl(=O)(=O)=O

color

APHA: ≤10

bp

198.7 °C/1013 hPa

mp

-18 °C

density

1.68 g/cm3 at 20 °C

storage temp.

15-25°C

InChI

1S/ClHO4/c2-1(3,4)5/h(H,2,3,4,5)

InChI key

VLTRZXGMWDSKGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Development and validation of an HPLC method for the determination of vancomycin in human plasma and its comparison with an immunoassay (PETINIA).: This study developed and validated an HPLC method using 70% perchloric acid for the determination of vancomycin in human plasma, highlighting its application in analytical chemistry for therapeutic drug monitoring (Usman and Hempel, 2016).
  • Determination of sunscreen agents in cosmetic products by reversed-phase high performance liquid chromatography.: The research utilized 70% perchloric acid in the sample preparation process, demonstrating its efficacy in the analysis of cosmetic products for regulatory and quality control purposes (Yao et al., 1998).
  • Colorimetric determination of hydroxyurea in human serum using high-performance liquid chromatography.: This article describes a colorimetric method involving 70% perchloric acid for sample preparation, which is crucial in clinical chemistry for monitoring hydroxyurea levels in patients (Manouilov et al., 1998).
  • Application of biological samples treated by wet-digested mineralization to cation-exchange high-performance liquid chromatography with post-column reaction by 4-(2-pyridylazo)-resorcinol.: This study details the use of 70% perchloric acid in the mineralization of biological samples, essential for accurate trace metal analysis in biological research (Sudo et al., 1991).
  • Colorimetric micromethods for glutaraldehyde determination by means of phenol and sulfuric acid or phenol and perchloric acid.: The research presents a micromethod utilizing 70% perchloric acid for the colorimetric determination of glutaraldehyde, applicable in biochemical and industrial laboratories for safety and quality assessments (Boratynski and Zal, 1990).

Analysis Note

Assay (acidimetric): 70.0 - 72.0 %
Color: ≤ 10 Hazen
Chlorate (ClO₃): ≤ 10 ppm
Chloride (Cl): ≤ 3 ppm
Free chlorine: ≤ 0.5 ppm
Phosphate and Silicate (as SiO₂): ≤ 5 ppm
Sulfate (SO₄): ≤ 10 ppm
Total nitrogen (N): ≤ 10 ppm
Heavy metals (as Pb): ≤ 1 ppm
Ag (Silver): ≤ 0.1 ppm
Al (Aluminium): ≤ 0.05 ppm
As (Arsenic): ≤ 0.05 ppm
Ba (Barium): ≤ 0.02 ppm
Be (Beryllium): ≤ 0.02 ppm
Bi (Bismuth): ≤ 0.1 ppm
Ca (Calcium): ≤ 0.5 ppm
Cd (Cadmium): ≤ 0.05 ppm
Co (Cobalt): ≤ 0.05 ppm
Cu (Copper): ≤ 0.1 ppm
Fe (Iron): ≤ 1.0 ppm
Ge (Germanium): ≤ 0.05 ppm
Hg (Mercury): ≤ 0.005 ppm
K (Potassium): ≤ 0.1 ppm
Li (Lithium): ≤ 0.02 ppm
Mg (Magnesium): ≤ 0.5 ppm
Mn (Manganese): ≤ 0.02 ppm
Mo (Molybdenum): ≤ 0.05 ppm
Na (Sodium): ≤ 0.5 ppm
Ni (Nickel): ≤ 0.1 ppm
Pb (Lead): ≤ 0.05 ppm
Sr (Strontium): ≤ 0.02 ppm
Ti (Titanium): ≤ 0.1 ppm
Tl (Thallium): ≤ 0.05 ppm
V (Vanadium): ≤ 0.05 ppm
Zn (Zinc): ≤ 0.1 ppm
Zr (Zirconium): ≤ 0.1 ppm
Residue on ignition (as sulfate): ≤ 30 ppm
Identity: passes test
Density (d 20/20): approx. 1.7
Conform to ACS, ISO, Reag.Ph Eur.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 1 - Skin Corr. 1A - STOT RE 2

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service