Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.00400

Supelco

Molybdic acid

about 85% MoO3, containing ammonium molybdate, EMPLURA®

Synonym(s):

Molybdic acid about 85% MoO3

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$609.00

$609.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
$609.00

About This Item

CAS Number:
Molecular Weight:
161.95
MDL number:
UNSPSC Code:
12352301
EC Index Number:
231-970-5
NACRES:
NA.21

$609.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Molybdic acid about 85% MoO3, (containing ammonium molybdate) EMPLURA®

Quality Level

product line

EMPLURA®

Assay

≥85% (precipitation titration)

form

solid

potency

2689 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

mp

300 °C

solubility

water: insoluble at 20 °C

density

3.1 g/cm3

bulk density

980 kg/m3

anion traces

chloride (Cl-): ≤0.01%
phosphate, silicate, arsenate (as PO43-): ≤0.005%
sulfate (SO42-): ≤0.02%

cation traces

Fe: ≤0.005%

storage temp.

2-30°C

SMILES string

[Mo](=O)(=O)(O)O

InChI

1S/Mo.2H2O.2O/h;2*1H2;;/q+2;;;;/p-2

InChI key

VLAPMBHFAWRUQP-UHFFFAOYSA-L

General description

(containing ammonium molybdate) EMPLURA®

Analysis Note

Assay (precipitative titration)≥ 85 %Chloride (Cl)≤ 0.01 %Phosphate, Arsenate, Silicate (as PO₄)≤ 0.005 %Sulfate (SO₄)≤ 0.02 %Fe (Iron)≤ 0.005 %NH₄ (Ammonium)about 6

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service