Skip to Content
MilliporeSigma

1.00164

SAFC

Boric acid

cryst., EMPROVE® API, Ph. Eur., BP, NF

Pharma Manufacturing

Synonym(s):

Orthoboric acid, Trihydroxidoboron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3BO3
CAS Number:
Molecular Weight:
61.83
MDL number:
UNSPSC Code:
12352106
EC Index Number:
233-139-2
Pricing and availability is not currently available.

Agency

BP
NF
Ph. Eur.

Quality Level

vapor pressure

2.6 mmHg ( 20 °C)

product line

EMPROVE® API

Assay

99.5-100.5% (alkalimetric)

form

solid

potency

2660 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

pH

3.8-4.8 (20 °C, 33 g/L in H2O)

mp

160 °C (dec.) (lit.)

solubility

49.2 g/L

density

1.489 g/cm3 at 23 °C

bulk density

400‑600 kg/m3

anion traces

sulfate (SO42-): ≤0.04%

cation traces

As: ≤0.0001%
heavy metals (as Pb): ≤0.0015%

application(s)

liquid formulation
ophthalmics
parenterals
pharmaceutical
semi-solid formulation

storage temp.

2-25°C

SMILES string

OB(O)O

InChI

1S/BH3O3/c2-1(3)4/h2-4H

InChI key

KGBXLFKZBHKPEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We provide industry-leading production know-how and comprehensive regulatory support for high-quality APIs, backed by nearly 200 years of experience. From a complete portfolio of innovative folates to custom synthesis, our customized and scalable solutions meet your specific production needs. As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Boric acid as an active ingredient is used in ophthalmic drugs, as antiseptic for cuts and burns, as well as antibacterial for dermatological treatments such as acne and athlete foot. Boric acid Emprove® API is manufactured according to ICH Q7 guidelines. A CEP and an US-DMF are available.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

API Information Package

Free of charge - Sign in to download

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service