Skip to Content
MilliporeSigma

1.00090

SAFC

Adipic acid

EMPROVE® ESSENTIAL, Ph. Eur., NF

Pharma Manufacturing

Synonym(s):

Adipic acid, Hexanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O4
CAS Number:
Molecular Weight:
146.14
MDL number:
UNSPSC Code:
12352106
E Number:
E355
EC Index Number:
204-673-3

Agency

NF
Ph. Eur.

Quality Level

reg. compliance

FCC

vapor pressure

0.097 hPa ( 18.5 °C)

description

EMPROVE® ESSENTIAL Ph Eur,NF,FCC,E 355

product line

EMPROVE® ESSENTIAL

form

solid

autoignition temp.

405 °C

potency

5700 mg/kg LD50, oral (Rat)
>7940 mg/kg LD50, skin (Rabbit)

technique(s)

API processing | cocrystal formation: suitable
API processing | salt formation: suitable

pH

2.7 (25 °C, 23 g/L in H2O)

mp

150.9 °C

transition temp

flash point 196 °C

solubility

15 g/L

density

1.36 g/cm3 at 25 °C

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

15-25°C

InChI

1S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)

InChI key

WNLRTRBMVRJNCN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Adipic acid is typically used as a pH modifier in pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

384.8 °F - closed cup

Flash Point(C)

196 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service