Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.00060

Millipore

CAYE Broth

Modified acc. to EVANS

Synonym(s):

CAYE Broth Modified Acc. To EVANS, Casein Hydrolysate Yeast Extract Broth

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$90.70

$90.70


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
100 G
$90.70

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

$90.70


Please contact Customer Service for Availability

Request more information

Quality Level

Sterilization Compatibility

autoclavable compatible

form

powder (dehydrated (DCM))

technique(s)

microbiological culture: suitable

pH

8.5-8.7 (25 °C)

solubility

39.75 g/L

application(s)

food and beverages
pathogen testing
pharmaceutical

storage temp.

15-25°C

suitability

selective for bacteria: Escherichia E. coli

General description

CAYE Broth is designed for the culture of pathogenic E. coli to enhance verotoxin production during growth.
The presence of casamino acids and yeast extract promote the growth of verotoxin producing E. coli. The presence of trace elements and resulting high pH value additionally supports the growth and toxin production.

Application

CAYE Broth is designed for the culture of pathogenic E. coli to enhance verotoxin production during growth in test samples.

Analysis Note

Appearance (clearness): clear
Appearance (colour): brown
pH-value (25 °C): 8.3 - 8.7
Growth (E. coli O157:H7 ATCC 43888): +
Duopath Verotoxin 1 (E. coli O157:H7 ATCC 43888): -
Duopath Verotoxin 2 (E. coli O157:H7 ATCC 43888): -
Growth (E. coli O157:H7 ATCC 43890): +
Duopath Verotoxin 1 (E. coli O157:H7 ATCC 43890): +
Duopath Verotoxin 2 (E. coli O157:H7 ATCC 43890): -
Growth (E. coli O157:H7 ATCC 43889): +
Duopath Verotoxin 1 (E. coli O157:H7 ATCC 43889): -
Duopath Verotoxin 2 (E. coli O157:H7 ATCC 43889): +

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service