Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09-822

Sigma-Aldrich

Anti-KLF5 (Krüppel-like factor 5) Antibody

serum, from rabbit

Synonym(s):

BTE-binding protein 2, Basic transcription element-binding protein 2, Colon krueppel-like factor, GC box binding protein 2, GC-box-binding protein 2, Intestinal-enriched krueppel-like factor, Kruppel-like factor 5, Kruppel-like factor 5 (intestinal), Tra

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ChIP
WB
species reactivity:
mouse
technique(s):
ChIP: suitable
western blot: suitable
citations:
3

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

technique(s)

ChIP: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KLF5(688)

General description

Krüppel-like factors (KLFs) are zinc finger-containing transcription factors with regulatory functions in cell growth, proliferation, differentiation, and embryogenesis. KLF5, (also known as BTEB2 and IKLF), is essential for mouse embryo development. KLF5 activates many genes during cardiovascular remodeling, including PDGFa/b, Egr-1, PAI-1, iNOS, and VEGF receptors, making KLF a potential therapeutic target for cardiovascular disease. KLF5 also plays a role in angiogenesis and in development of several tumor types including breast, prostate, bladder, colon, esophagus, and skin. KLF5 is closely related to other of the KLF family members. However, KLF5 and KLF4 exhibit opposite effects on regulation of gene transcription, despite binding to similar, if not identical, cis-acting DNA sequences. In addition, KLF4 and 5 exert contrasting effects on cell proliferation in many instances; while KLF4 is an inhibitor of cell growth, KLF5 stimulates proliferation.

Specificity

This antibody recognizes Krüppel-like factor 5 at the N-terminus.

Immunogen

Epitope: N-terminus
Recombinant protein corresponding to mouse Krüppel-like factor 5 at the N-terminus.

Application

Anti-KLF5 (Krüppel-like factor 5) Antibody is a Rabbit Polyclonal Antibody for detection of KLF5 (Krüppel-like factor 5) also known as Krüppel-like factor 5 & has been validated in WB.
Chromatin Immunoprecipitation Analysis (ChIP):
A representative lot was used by an independent laboratory in ChIP. (Ng Huck Hui, Genome Institute of Singapore)
Research Category
Epigenetics & Nuclear Function

Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Pluripotent & Early Differentiation

Quality

Evaluated by Western Blot in mouse embryonic stem cell chromatin extract.

Western Blot Analysis: 1:2,000 dilution of this antibody detected Krüppel-like factor 5 on 10 µg of mouse embryonic stem cell chromatin extract.

Target description

~55 kDa observed. An uncharacterized band appears at ~30 kDa

Physical form

Rabbit polyclonal serum containing 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Mouse embryonic stem cell chromatin extract

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chengli Dou et al.
Journal of molecular cell biology, 15(7) (2023-08-03)
The effective proliferation and differentiation of trophoblast stem cells (TSCs) is indispensable for the development of the placenta, which is the key to maintaining normal fetal growth during pregnancy. Kruppel-like factor 5 (Klf5) is implicated in the activation of pluripotency
Zhenyu Yang et al.
Oncogene, 43(23), 1779-1795 (2024-04-23)
Transcription factors (TFs) engage in various cellular essential processes including differentiation, growth and migration. However, the master TF involved in distant metastasis of nasopharyngeal carcinoma (NPC) remains largely unclear. Here we show that KLF5 regulates actin remodeling to enhance NPC
Dong Han et al.
Science advances, 8(7), eabe4375-eabe4375 (2022-02-17)
Oct4 collaborates primarily with other transcriptional factors or coregulators to maintain pluripotency. However, how Oct4 exerts its function is still unclear. Here, we show that the Oct4 linker interface mediates competing yet balanced Oct4 protein interactions that are crucial for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service