Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

09-158

Sigma-Aldrich

Anti-ALK7 Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$517.00

$517.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1384

Request a Bulk Order

Select a Size

Change View
200 μL
$517.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$517.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1384

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

ALK7 (ACVR1C activin A receptor, type IC; ACVRLK4) is a transmembrane type IC receptor serine kinase for activin or activin-like ligands. Activins, are dimeric factors that belong to the TGF-superfamily of signaling proteins. Activins signal through the hetrodimeric complex that includes two type I (I and IB) and two type II (II and IIB) receptors. The type I receptors (i.e. ALK7) contains a Ser/Thr kinase domain in its cytoplasmic domain that is essential for signaling, while the type II are receptors are essential for ligand binding (Harrison, 2004 & Gray, 2000). Receptor activation involves ligand binding to the promiscuous type II receptor that is known to bind to many ligands. This leads to the recruitment of ALK7 and the formation of a stable heterodimer that results in the phosphorylation and activation of ALK7.

Specificity

Recognizes ALK7

Immunogen

Synthetic peptide designed against the N-terminal region of human ALK7

Application

Anti-ALK7 Antibody is an antibody against ALK7 for use in WB.
Research Category
Signaling
Research Sub Category
Cytokines & Cytokine Receptors

Developmental Signaling

Growth Factors & Receptors

Quality

Routinely evaluated by western blot on either human or rat brain lysate

Target description

55 kDa

Physical form

Affinity purified rabbit polyclonal IgG in storage buffer containing PBS with 0.05% sodium azide.
Affinity purified using immunogen

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alan Jung Park et al.
eLife, 6 (2017-09-21)
Long-lasting forms of synaptic plasticity and memory require de novo protein synthesis. Yet, how learning triggers this process to form memory is unclear. Translin/trax is a candidate to drive this learning-induced memory mechanism by suppressing microRNA-mediated translational silencing at activated
Nicole M Sayles et al.
Cell reports, 38(10), 110475-110475 (2022-03-10)
Mitochondrial cardiomyopathies are fatal diseases, with no effective treatment. Alterations of heart mitochondrial function activate the mitochondrial integrated stress response (ISRmt), a transcriptional program affecting cell metabolism, mitochondrial biogenesis, and proteostasis. In humans, mutations in CHCHD10, a mitochondrial protein with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service