Skip to Content
MilliporeSigma
All Photos(1)

Documents

07-877

Sigma-Aldrich

Anti-phospho-PKCη (Thr655) Antibody

Upstate®, from rabbit

Synonym(s):

PRKCH, PKCeta, PKCh, protein kinase C, eta, KPCL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pThr655)

Gene Information

human ... PRKCH(5583)

Specificity

Predicted to cross-react with Mouse and Rat (100% homologous) based on sequence homology.
Recognizes phosphorylated PKCη (Thr655). Not expected to cross-react with other PKC isoforms

Immunogen

Peptide corresponding to amino acid region encompassing the human, mouse, and rat phospho-PKCη (Thr655).

Application

Immunoblot Analysis: A 0.50 μg/mL dilution of this lot detected phospho-PKCη (Thr655) in RIPA lysate of PMA treated Jurkat cells.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling
This Anti-phospho-PKCη (Thr655) Antibody is validated for use in WB for the detection of phospho-PKCη (Thr655).

Quality

Routinely evaluated by immunoblot.

Target description

~80 kDa

Physical form

100 μL of affinity purified rabbit polyclonal IgG in 50% storage buffer (PBS (without Mg2+ and Ca2+), pH 7.3 containing 1.0 mg/mL BSA (IgG, protease free) and 0.05% sodium azide).
Affinity purified

Storage and Stability

Stable for 2 years at -80°C from date of shipment. Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -80°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keisuke Taniuchi et al.
PloS one, 7(4), e35674-e35674 (2012-04-26)
A novel function for the binder of Arl two (BART) molecule in pancreatic cancer cells is reported. BART inhibits invasiveness of pancreatic cancer cells through binding to a Ca(2+)-dependent, phosphorylated, guanosine triphosphatase (GTPase) membrane fusion protein, annexin7 (ANX7). A tumor
Peter Gruber et al.
Journal of biochemistry, 149(3), 331-336 (2010-12-28)
Protein kinase C (PKC) is a family of at least 10 isozymes involved in the activation of different signal transduction pathways. The exact function of these isozymes is not known at present. Isozyme-selective inhibitors would be important to explain the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service