Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07-294-I

Sigma-Aldrich

Anti-phospho-GluR2 (Ser880)

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$242.00
100 μG
$619.00

$242.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$242.00
100 μG
$619.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$242.00


Please contact Customer Service for Availability

Request a Bulk Order

antibody form

purified antibody

clone

polyclonal

purified by

affinity chromatography

species reactivity

rabbit, rat

concentration

(Please refer to lot specific datasheet.)

technique(s)

western blot: suitable

isotype

IgG

UniProt accession no.

General description

KLH-conjugated Linear Peptide corresponding to 9 amino acids surrounding phosphoserine 880 from the the C-terminal region of Rat Glutamate receptor 2.

Specificity

Anti-phospho-GluR2 (Ser880) is a rabbit polyclonal antibody that detects glutamate receptor 2 phosphorylated on serine 880.

Application

Epitope: C-terminus

Quality

Evaluated by Western Blotting in Rat Brain Microsomal Prep with Lambda phosphatase treatment.

Western Blotting Analysis: 2 µg/mL of this antibody detected Glutamate receptor 2 phosphorylated on Serine 880 in untreated Rat brain microsomal preparations, but not in preparations treated with Lambda phosphatase.

Physical properties

~110 kDa observed; 98.69 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Rat. Predicted to react with Human, Mouse, Monkey, and Chimpanzee based on 100% sequence homology.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service