Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

07-217-I

Sigma-Aldrich

Anti-NAG-1/GDF15

from rabbit, purified by affinity chromatography

Synonym(s):

Macrophage inhibitory cytokine 1, MIC-1, NSAID-activated gene 1 protein, NAG-1, NSAID-regulated gene 1 protein, NRG-1, Placental TGF-beta, Placental bone morphogenetic protein, Prostate differentiation factor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$593.00

$593.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$593.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$593.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

General description

Growth/differentiation factor 15 (UniProt: Q99988; also known as GDF-15, Macrophage inhibitory cytokine 1, MIC-1, NSAID-activated gene 1 protein, NAG-1, NSAID-regulated gene 1 protein, NRG-1, Placental TGF-beta, Placental bone morphogenetic protein, and Prostate differentiation factor) is encoded by the GDF15 (also known as MIC1, PDF, PLAB, PTGFB) gene (Gene ID: 9518) in human. NAG-1 is a member of the TGF-beta super family, however, it does not display strong homology to existing families. NAG-1 is synthesized as a 308-amino acid pro-NAG-1 monomer and then dimerizes by a specific disulfide linkage. The pro-NAG-1 dimer is then cleaved by furin-like proteases at an RXXR site, forming a 112 amino acid C-terminal dimeric protein and pro-peptide. It is expressed in placenta and is found in lower levels in prostate, kidney, and colon. Its expression is significantly increased during inflammation, injury, and malignancy. Increase of NAG-1 expression is a feature of many cancers including breast, colon, pancreas and prostate. NAG-1 expression is also shown to increase following exposure to a number of non-steroidal anti-inflammatory drugs. Induced expression of NAG-1 is shown to result in stimulation of apoptosis and inhibition of cell growth.

Specificity

This polyclonal antibody specifically detects Growth/differentiation factor 15 in human.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal end of human NAG-1/GDF15.

Application

Detect Growth/differentiation factor 15 using this rabbit polyclonal Anti-NAG-1/GDF15, Cat. No. 07-217-I, useing Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Signaling
Western Blotting Analysis: 2 µg/mL from a representative lot detected NAG-1/GDF15 in 10 µg of HepG2 conditioned media.



Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected NAG-1/GDF15 in human placenta tissue.


Quality

Evaluated by Western Blotting in LNCap Conditioned Media.

Western Blotting Analysis: 2 µg/mL of this antibody detected NAG-1/GDF15 in 10 µg of LNCap Conditioned Media.

Target description

34.14 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: 07-217

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service