Skip to Content
MilliporeSigma

Skip To

07-2139

Anti-Cysteine Sulfenic Acid Antibody

serum, from rabbit

Sign In to View Organizational & Contract Pricing.

About This Item

eCl@ss:
32160702
UNSPSC Code:
12352203
Clone:
polyclonal
Species reactivity:
mouse, rat, human
Application:
WB
Citations:
2


biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

species reactivity (predicted by homology)

mammals

technique(s)

western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Multiple kDa are observed that increases with oxidized stress (H2O2).
Protein sulfenic acid formation is a reversible post-translational modification that may be used to monitor protein oxidation on reactive cysteines within target proteins. This can be detected with protein sulfenic acid derivatised with dimedone.

Immunogen

Epitope: Other
KLH-conjugated corresponding to cysteine sulfenic acid.

Application

Anti-Cysteine Sulfenic Acid detects levels of Cysteine Sulfenic Acid proteins & has been published & validated for use in WB.
Research Category
Apoptosis & Cancer
Research Sub Category
General Post-translation Modification
Western Blot Analysis: A previous lot of this antibody detected cysteine sulfenic acid in NIH/3T3 and HEK293 cell lysates.

Biochem/physiol Actions

This antibody recognizes proteins containing cysteine sulfenic acid derivitized with dimedone.

Physical form

Unpurified
Unpurified rabbit polyclonal with 0.05% sodium azide.

Preparation Note

Stable for 1 year at from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Rat ventricular myocyte lysate
Evaluated by Western Blot in rat ventricular myocyte lysate.

Western Blot Analysis: 1:10,000 dilution of this antibody detected cysteine sulfenic acid on 10 µg of rat ventricular myocyte lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters