Skip to Content
MilliporeSigma
All Photos(1)

Documents

07-2101-M

Sigma-Aldrich

Anti-phospho PIX Antibody (Ser340)

from rabbit, purified by affinity chromatography

Synonym(s):

Rho guanine nucleotide exchange factor (GEF) 7, PAK-interacting exchange factor beta, guanine nucleotide exchange factor 7, rho guanine nucleotide exchange factor 7, SH3 domain-containing proline-rich protein, p85, PAK3, Beta-Pix, P85COOL1, COOL-1, P85,

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology), human (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

phosphorylation (pSer340)

Gene Information

human ... PAK3(5063)

General description

p-21-activated kinase (PAK) interacting exchange factor (PIX) is a guanine nucleotide exchange factor. PIX is a necessary component for the recruitment, regulation and activation of Rac1, as it is able to directly bind Rac1 via its SH3 domain. Studies indicate that PIX may also have an active role as a positive regulator of apoptosis, an inducer of membrane ruffling and in cell migration. Along with Rac1, PIX has been shown to regulate and interact with GIT1 and NOD2. Alternative splicing and initiation give rise to 8 known isoforms. Expression of PIX has been localized to the central nervous system. Isoforms B, C and E have highest expression in the testis and brain.

Specificity

This antibody recognizes PIX phosphorylated at Ser340.

Immunogen

Epitope: Phosphorylated Ser340
KLH-conjugated linear peptide corresponding to mouse PIX phosphorylated at Ser340.

Application

This phospho PIX antibody is validated for use in WB for the detection of the phospho PIX protein.

Quality

Evaluated by Western Blot in PIX transfected cell lysates.

Western Blot Analysis: A 1:1000 dilution of this antibody detected PIX on 10 µg of Pix transfected cell lysates.

Target description

~ 75 kDa

Physical form

Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide, 1 mg/mL BSA and 30% glycerol.

Analysis Note

Control
PIX transfected cell lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service