Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

07-1490-M

Sigma-Aldrich

Anti-Calcineurin Antibody

from rabbit, purified by affinity chromatography

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

mammals

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... PPP3CA(5530)

General description

Calcineurin/PP2B is a serine/threonine protein phosphatase involved in the mediation of signal tranduction in a wide variety of cellular responses. Calcineurin/PP2B has been linked to Pmk1 MAP kinase and PI pathways, as well as to members of the small GTPase Rab/Ypt family and Type II myosin. Furthermore, calcineurin participates in the transduction of extracellular signals to the nucleus by dephosphorylation of members of the NFAT family of transcription factors. These linkages demonstrate calcineurin/PP2B′s multifunctional capabilities in the regulation of calcium signaling.

Specificity

Broad reactivity with mammal species is expected.
Recognizes both the catalytic A and regulatory B subunits of calcineurin, a calcium- and calmodulin-dependent protein phosphatase.

Immunogen

Purified bovine brain calcineurin.

Application

Detect Calcineurin using this Anti-Calcineurin Antibody validated for use in WB, IH(P) & IP.
Immunohistochemisty (paraffin): Representative testing from a previous lot. Calcineurin representative staining pattern/morphology in human vascular tissue. Tissue was pretreated with citrate pH 6, antigen retrieval. A previous lot of this antibody was diluted to 1:100, using IHC-Select Detection with HRP-DAB.

Immunoprecipitation: 3 μL per reaction of a previous lot of this antibody was used in Immunoprecipitation.

Optimal working dilutions must be determined by end user
Research Category
Signaling
Research Sub Category
GPCR, cAMP/cGMP & Calcium Signaling

Quality

Routinely evaluated by Western Blot on PC3 lysates.

Western Blot Analysis:
1:500 dilution of this lot detected PP2B B on 10 μg of PC3 lysates.

Target description

19 kDa

Linkage

Replaces: AB1694

Physical form

Affinity purified
Purified rabbit polyclonal IgG in buffer containing TBS and 40% glycerol as a cryopreservative.

Storage and Stability

Stable for 1 year at -20°C in undiluted aliquots from date of receipt; at this temperature the solution will remain unfrozen.
Handling Recommendations: Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerolcontaining solutions to become frozen during storage. Note: Variability in freezer temperatures below -20°C may cause glycerolcontaining solutions to become frozen during storage.

Analysis Note

Control
PC3 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcineurin triggers reactive/inflammatory processes in astrocytes and is upregulated in aging and Alzheimer's models.
Norris, Christopher M, et al.
The Journal of Neuroscience, 25, 4649-4658 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service