Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07-100

Sigma-Aldrich

Anti-Nckβ Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
$439.00

$439.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
200 μG
$439.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$439.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NCK2(8440)

Specificity

Nckβ

Immunogen

peptide corresponding to amino acids 175-190 of murine Nckβ (EAPSFLSLRRGTALSN)

Application

Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling
This Anti-Nckβ Antibody is validated for use in IP, WB for the detection of Nckβ.

Quality

routinely evaluated by immunoblot on RIPA lysates of murine testes and spleen homogenates

Target description

47kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Format: Purified
Protein A chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Ling et al.
The Journal of biological chemistry, 276(22), 18908-18914 (2001-03-30)
Hematopoietic progenitor kinase 1 (HPK1), a mammalian Ste20-related serine/threonine protein kinase, is a hematopoietic-specific upstream activator of the c-Jun N-terminal kinase. Here, we provide evidence to demonstrate the involvement of HPK1 in T cell receptor (TCR) signaling. HPK1 was activated
Matthew Oser et al.
Journal of cell science, 123(Pt 21), 3662-3673 (2010-10-26)
Invadopodia are matrix-degrading membrane protrusions in invasive carcinoma cells enriched in proteins that regulate actin polymerization. The on-off regulatory switch that initiates actin polymerization in invadopodia requires phosphorylation of tyrosine residues 421, 466, and 482 on cortactin. However, it is
Lisa Buvall et al.
Nature communications, 4, 2863-2863 (2013-11-30)
The ubiquitously expressed adapter proteins Nck1/2 interact with a multitude of effector molecules to regulate diverse cellular functions including cytoskeletal dynamics. Here we show that Nck1, but not Nck2, is a substrate of c-Cbl-mediated ubiquitination. We uncover lysine 178 in
Derek L Clouthier et al.
Molecular and cellular biology, 35(9), 1573-1587 (2015-02-19)
Development of the cardiovascular system is critically dependent on the ability of endothelial cells (ECs) to reorganize their intracellular actin architecture to facilitate migration, adhesion, and morphogenesis. Nck family cytoskeletal adaptors function as key mediators of actin dynamics in numerous
Barbara Calabrese et al.
Nature communications, 13(1), 6037-6037 (2022-10-14)
During early ischemic brain injury, glutamate receptor hyperactivation mediates neuronal death via osmotic cell swelling. Here we show that ischemia and excess NMDA receptor activation cause actin to rapidly and extensively reorganize within the somatodendritic compartment. Normally, F-actin is concentrated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service