Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06-680-AF647

Sigma-Aldrich

Anti-Androgen Receptor Antibody, Alexa Fluor 647 Conjugate

from rabbit, ALEXA FLUOR 647

Synonym(s):

Androgen receptor, Dihydrotestosterone receptor, Nuclear receptor subfamily 3 group C member 4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$560.00

$560.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$560.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

$560.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 647

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human

species reactivity (predicted by homology)

canine (based on 100% sequence homology), bovine (based on 100% sequence homology), mouse (based on 100% sequence homology), horse (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

bovine ... Ar(280675)
dog ... Ar(403588)
human ... AR(367)
mouse ... Ar(11835)
rat ... Ar(24208)

General description

Androgen receptor (UniProt P15207; also known as Dihydrotestosterone receptor, Nuclear receptor subfamily 3 group C member 4) is encoded by the Ar (also known as Andr, Nr3c4, Tfm) gene (Gene ID 24208) in rat species. Androgen receptor (AR) is a hormone-activated transcription regulator that plays important roles in normal prostate development and function, as well as in malignant prostate transformation. The binding of an androgenic hormone (testosterone or dihydrotestosterone) induces a conformational change in AR, resulting its dissociation from heat schock proteins and subsequent translocation from the cytosol to nucleus, where it dimerizes and mediates either transcription activation or repression in a target gene-specific manner. The androgen insensitivity syndrome (AIS) or testicular feminization (TFM) is caused by a mutation of the androgen receptor gene located on the X chromosome.

Immunogen

Epitope: This antibody recognizes the Modulating Region within the N-terminal of androgen receptor.
KLH-conjugated linear peptide corresponding to the N-terminal sequence of rat androgen receptor.

Application

Detect Androgen Receptor using this Anti-Androgen Receptor Antibody, Alexa Fluor 647 Conjugate validated for use in Immunocytochemistry.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Hormones & Receptors
The unconjugated antibody (Cat. No. 06-680) is shown to be suitable also for chromatin immunoprecipitation (ChIP), ChIP-seq, Western blotting, immunoprecipitation, flow cytometry, immunohistochemistry applications.

Quality

Evaluated by Immunocytochemistry in MCF-7 cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Androgen Receptor in MCF-7 cells.

Target description

~110 kDa observed.

Physical form

Affinity purified
Purified rabbit polyclonal antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service