Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

06-1372

Sigma-Aldrich

Anti-acetyl (Lys26) phospho (Ser27) Histone H1.4 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H1.4, Histone H1b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rhesus macaque (based on 100% sequence homology), canine (based on 100% sequence homology), orangutan (based on 100% sequence homology), giant panda (based on 100% sequence homology)

technique(s)

dot blot: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pLys26)

Gene Information

dog ... H1-4(488269)
human ... H1-4(3008)
rhesus monkey ... H1-4(696872)

General description

H1.4 is one of four variants of histone H1 (H1.2-H1.5) that is found in somatic cells. The H1 histones may function as linker proteins binding DNA via their GH domain, and enabling the wrapping of DNA around the core nucleosomal structure formed from the association of H2A, H2B, H3, and H4. In response to various stimuli, histones undergo a number of post-translational modifications (PTM) including acetylation, phoshorylation, and methylation. Previous studies have suggested that H1.4 may be phosphorylated during mitosis and interphase resulting in increased transcription of target genes by RNA polymerase I and II, and therefore this PTM on H1.4 may have implications for cell growth. H1.4 may also undergo acetylation of its N-terminal lysine residues. Often, it is the pattern of different modifications on several histone residues that alter chromatin and affect gene expression, rather than modification of single residues.

Specificity

Other homologies: Rat and Mouse (82% sequence homology).
This antibody recognizes the N-terminus of Histone H1.4 acetylated at Lys26 and phosphorylated at Ser27.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H1.4 acetylated at Lys26 and phosphorylated at Ser27.

Application

Anti-acetyl (Lys26) phospho (Ser27) Histone H1.4 Antibody is a rabbit polyclonal antibody for detection of acetyl (Lys26) phospho (Ser27) Histone H1.4 also known as Histone H1.4, Histone H1b & has been validated in WB, DB.
Dot Blot Analysis: A 1:1,320 dilution from representative lot detected acetyl (Lys26) phospho (Ser27) Histone H1.4 peptides.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Evaluated by Western Blot in colcemid treated HeLa acid extract.

Western Blot Analysis: A 1:3,000 dilution of this antibody detected acetyl (Lys26) phospho (Ser27) Histone H1.4 in 10 µg of colcemid treated HeLa acid extract.

Target description

~35 kDa observed. Uncharacterized bands may be observed at ~70 kDa and ~17 kDa in some acid extracts.

Physical form

Affinity purified
Purified rabbit polyclonal serum in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Colcemid treated HeLa acid extract

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean-Michel Terme et al.
FEBS letters, 588(14), 2353-2362 (2014-05-31)
In mammals, the linker histone H1, involved in DNA packaging into chromatin, is represented by a family of variants. H1 tails undergo post-translational modifications (PTMs) that can be detected by mass spectrometry. We developed antibodies to analyze several of these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service