Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

06-1364

Sigma-Aldrich

Anti-SRp30b (SC35) Antibody

from rabbit

Synonym(s):

serine/arginine-rich splicing factor 2, Splicing factor SC35, Splicing component, 35 kDa, splicing factor, arginine/serine-rich 2, SR splicing factor 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SRSF2(6427)

General description

SRp30b, also known as SC35, and Splicing factor, arginine/serine-rich 2 (SFRS2) is an essential splicing factor. This protein is encoded by the SFRS2 gene and is a member of the splicing factor SR family. SRp30b is involved in pre-mRNA splicing and is found in bodies in the nucleus that are highly enriched in poly(A) RNA. These nuclear bodies are referred to as speckles, SC 35 domains, or splicing factor compartments (SFCs). The purpose of these bodies is not completely characterized. Speckles correspond largely to, if not entirely to, ultra structures called inter-chromatin granule clusters (IGCs). SRp30b is required for pre-mRNA splicing events and is highly involved in spliceosomal interaction and assembly. This SR protein is necessary for assembly of the ATP-dependent splicing complex. It is also necessary for the ATP-dependent communication between pre-mRNA and U1 and U2 snRNPs.

Immunogen

Linear peptide corresponding to human SRp30b.

Application

Anti-SRp30b (SC35) Antibody is a Rabbit Polyclonal Antibody for detection of SRp30b (SC35) also known as serine/arginine-rich splicing factor 2, Splicing factor SC35 & has been validated in WB, ICC.
Western Blot Analysis:
A 1:500 dilution of a representative lot of this antibody detected SRp30b in 10 µg of HeLa cell lysate.

Immunoprecipitation Analysis:
A representative lot of this antibody was used by an independent laboratory in IP. (Jiang, K., et al. (2009). Endocrinology. 150(5):2087-2097.)

Quality

Evaluated by Immunocytochemistry Analysis:
A 1:500 dilution of this antibody detected SRp30b in NIH/3T3 cells.

Target description

~35 kDa observed. Uncharacterized bands may be observed at ~10, 58, and 78 kDa

Physical form

Format: Purified

Analysis Note

Control
NIH/3T3 cells.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service