Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-878

Sigma-Aldrich

Anti-Protein C Antibody, clone GMA-093

clone GMA-093, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$466.00

$466.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$466.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$466.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

GMA-093, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PROC(5624)

Specificity

Protein C

Immunogen

Purified human Protein C

Application

Anti-Protein C Antibody, clone GMA-093 detects level of Protein C & has been published & validated for use in ELISA & WB.
Research Category
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms

Quality

routinely evaluated by immunoblot on human Protein C

Target description

non-reduced, Mr 62kDa, reduced , Mr 41kDa

Physical form

DEAE purified
Format: Purified

Storage and Stability

stable 2 years at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandeep Kumar et al.
Frontiers in neuroscience, 14, 554094-554094 (2020-10-20)
The organization of the anterior forebrain pathway (AFP) of songbirds important for context-dependent singing is similar to that of cortical basal ganglia loops (CBG) in mammals, which underlie motor behaviors including vocalization. Since different components of the AFP express high

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service