Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

05-843R

Sigma-Aldrich

Anti-phospho-Bcl-2 (Ser70) Antibody, clone 69-10C-2-10C-18, rabbit monoclonal

clone 69-10C-2-10C-18, from rabbit, unconjugated

Synonym(s):

B-cell CLL/lymphoma 2, B-cell lymphoma protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$436.00

$436.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$436.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$436.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

69-10C-2-10C-18, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer70)

Gene Information

human ... BCL2(596)

General description

The Bcl name is derived from B-cell chronic lymphocytic leukemia, and was given to genes involved in that disease. Members of the Bcl-2 family share homologous domains (BH) through which they are able to homo- and hetero-dimerize. Bcl-2 plays an important role in cytochrome c release from the mitochondria; it also binds Apaf1, inhibiting Apaf-1 association with caspase-9. Bcl-2 overexpression in indicated in a number of human cancers, including melanoma, breast, prostate, and lung, and it correlates with resistance to cancer treatment (presumably due to decreased apoptosis). Bcl-2 has also been implicated in schizophrenia and autoimmune disorders. Bcl-2 phosphorylated at Ser-70 plays a critial role in the translocation of Bax from the cytosol to the mitochondria, and this may regulate NO-induced neuronal apoptosis. The molecular mechanism of Bcl-2 phosphorylation and its relationship to Bax is largely unknown.

Specificity

This antibody recognizes phosphorylated Bcl-2 at Ser70.

Immunogen

Epitope: Ser70
KLH-conjugated linear peptide corresponding to human phosphorylated Bcl-2 at Ser70.

Application

Detect phospho-Bcl-2 (Ser70) using this Anti-phospho-Bcl-2 Antibody, clone 69-10C-2-10C-18 validated for use in WB, IP, FC.
Immunoprecipitation Analysis: 1ug from a previous lot immunoprecipitated phosphorylated Bcl-2 from 100 ug of jurkat lysate treated with paciltaxel. Arrow indicates phospho-Bcl-2 (~26kD).

Flow Cytometry Analysis: 0.1ug/ml from a previous lot was used to detect Bcl-2 in Jurkat cell lysate.
Research Category
Apoptosis & Cancer
Research Sub Category
BCL2 & Inhibition

Quality

Evaluated by Western Blot in paclitaxel treated and untreated Jurkat cell lysate.

Western Blot Analysis: 1:1,000 dilution of this antibody detected Bcl-2 on 10 µg of Jurkat cell lysate.

Target description

26 kDa

Physical form

Protein A purified
Purified rabbit monoclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide and 40% Glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Paclitaxel treated and untreated Jurkat cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Contribution of Bcl-2 phosphorylation to Bak binding and drug resistance.
Dai, Haiming, et al.
Cancer Research, 73, 6998-7008 (2013)
Michela Guglielmotto et al.
Autophagy, 10(10), 1827-1843 (2014-08-20)
The role of autophagy and its relationship with apoptosis in Alzheimer disease (AD) pathogenesis is poorly understood. Disruption of autophagy leads to buildup of incompletely digested substrates, amyloid-β (Aβ) peptide accumulation in vacuoles and cell death. Aβ, in turn, has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service