Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-827

Sigma-Aldrich

Anti-Glucocorticoid Receptor Antibody, clone FIGR

clone FIGR, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
FIGR, monoclonal
application:
IP
WB
species reactivity:
hamster, mouse, rat
technique(s):
immunoprecipitation (IP): suitable
western blot: suitable
citations:
4

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

FIGR, monoclonal

species reactivity

hamster, mouse, rat

should not react with

human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NR3C1(2908)

Specificity

glucocorticoid receptor

Immunogen

Native glucocorticoid receptor purified from rat liver

Application

Detect Glucocorticoid Receptor with Anti-Glucocorticoid Receptor Antibody, clone FIGR (Mouse Monoclonal Antibody), that has been shown to work in IP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

routinely evaluated by immunoblot on RIPA lysates from L6, PC12, and 3T3/A31 cells.

Target description

85-100kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Format: Purified
Protein G Purified

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R C Kaltreider et al.
Environmental health perspectives, 109(3), 245-251 (2001-05-03)
Chronic human exposure to nonovertly toxic doses of arsenic is associated with an increased risk of cancer. Although its carcinogenic mechanism is still unknown, arsenic does not directly cause DNA damage or mutations and is therefore thought to act principally
J E Bodwell et al.
The Journal of biological chemistry, 266(12), 7549-7555 (1991-04-25)
Glucocorticoid receptors in vivo are phosphorylated in the absence of hormone and become hyperphosphorylated in the presence of glucocorticoid agonist but not antagonists (Ortí, E., Mendel, D.B., Smith, L.I., and Munck, A. (1989) J. Biol. Chem. 264, 9728-9731). As a
J Richardson et al.
Molecular endocrinology (Baltimore, Md.), 13(4), 546-554 (1999-04-09)
The glutamate synthetase gene (GS) contains a composite glucocorticoid response element (cGRE) comprised of a GRE and an adjacent element with features of both a cAMP-response element (CRE) and a 12-O-tetradecanoylphorbol 13-acetate (TPA) response element (TRE). The CRE/TRE element of
Arman Nayebosadri et al.
American journal of physiology. Cell physiology, 305(3), C309-C322 (2013-05-25)
The lamina serves to maintain the nuclear structure and stiffness while acting as a scaffold for heterochromatin and many transcriptional proteins. Its role in endothelial mechanotransduction, specifically how nuclear mechanics impact gene regulation under shear stress, is not fully understood.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service