Skip to Content
MilliporeSigma
All Photos(2)

Documents

05-1587

Sigma-Aldrich

Anti-FOXO3a/FKHRL1 Antibody, clone 15F7.2

clone 15F7.2, from mouse

Synonym(s):

Forkhead box protein O3, AF6q21 protein, Forkhead in rhabdomyosarcoma-like 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

15F7.2, monoclonal

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FOXO3(2309)

General description

Forkhead box protein O3 (FOXO3a), or FKHRL1, is a ubiquitous transcription factor that triggers the expression of pro-apoptotic genes under conditions of oxidative stress. FOXO3a is regulated by the Akt kinase which phosphorylates FOXO3a on serine 253, 315, and on threonine 32 residues. Modified FOXO3a is recognized by the 14-3-3 proteins which export FOXO3a to the cytoplasm, and consequently terminates its transcriptional activity. FOXO3a may also be regulated by lysine methylation in neuronal cells.

Immunogen

GST-tagged recombinant protein corresponding to human FOX03a.

Application

Anti-FOXO3a/FKHRL1 Antibody, clone 15F7.2 is a Mouse Monoclonal Antibody for detection of FOXO3a/FKHRL1 also known as Forkhead box protein O3, AF6q21 protein, Forkhead in rhabdomyosarcoma-like 1 & has been validated in WB & IHC.
Immunohistochemistry Analysis: A 1:300 dilution from a representative lot detected FOXO3a/FKHRL1 in human thyroid gland tissue and human breast cancer tissue.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blot in rat liver tissue lysate.

Western Blot Analysis: 0.2 µg/mL of this antibody detected FOXO3a/FKHRL1 in 10 mg of rat liver tissue lysate.

Target description

~71 kDa observed. An uncharacterized band may be observed at ~28 kDa in some tissue lysates.

Linkage

Replaces: 04-1007

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat liver tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service