Skip to Content
MilliporeSigma
All Photos(1)

Documents

05-1100X

Sigma-Aldrich

Anti-Sulfotyrosine Antibody, Clone Sulfo-1C-A2

clone Sulfo-1C-A2, Upstate®, from mouse

Synonym(s):

Clone Sulfo-1C-A2 Antibody, Sulfotyrosine Detection Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

Sulfo-1C-A2, monoclonal

species reactivity

human, porcine, mouse, bovine

species reactivity (predicted by homology)

all

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

shipped in

wet ice

target post-translational modification

unmodified

General description

Sulfotyrosine is a common post-translational modification of secretory, plasma membrane, and lysosomal proteins. The sulfation reaction is catalyzed by specific sulfotransferase enzymes in the trans-Golgi-network. Tyrosine sulfation occurs in all multicellular organisms and up to 1% of the tyrosines in the total protein content of a cell can be sulfated. Sulfotyrosines mediates many receptor-ligand binding interactions, including the interaction of P-selectin glycoprotein ligand-1 with P-selectin, factor VIII binding to von Willebrand factor, and interactions between thrombin and hirudin. Sulfotyrosine is also involved in the entry of several parasites and viruses; for example, tyrosine sulfation of the HIV-1 co-receptor CCR5 is required for viral entry into host cells, and tyrosine sulfation of the Duffy antigen/receptor for chemokines is crucial for erythrocyte invasion by the malaria parasite Plasmodium vivax.
This clone was derived from the antibody described in Kehoe et al., 2006.

Specificity

Recognizes sulfated tyrosine residues on various proteins. Does not recognize phosphotyrosine or unmodified tyrosines.
The antibody is predicted to recognize any sulfotyrosine, regardless of surrounding amino acid sequence.

Immunogen

Epitope: Sulfated Tyrosine
This antibody was developed using a phage display library as described in Kehoe et al., 2006. The selection antigens were KAKISDP-DY(SO3)MTGYMDAC and KDKKYATEY(SO3)-EYLDYDFC

Application

Anti-Sulfotyrosine Antibody, Clone Sulfo-1C-A2 is a high quality Mouse Monoclonal Antibody for the detection of Sulfotyrosine & has been validated in WB, IP, ELISA.
Research Category
Signaling
Research Sub Category
General Post-translation Modification
WB, IP, ELISA

Quality

Assayed using sulfatase treated bovine fibrinogen vs. untreated fibrinogen.

Target description

n/a

Physical form

Culture supernatant, purified by protein G affinity chromatography
Format: Purified
Mouse IgG2aκ purified supernatant in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH 7.4 .

Storage and Stability

Stable for 1 year from date of receipt when stored at +2-8°C.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service