Skip to Content
MilliporeSigma
All Photos(1)

Documents

03-314-M

Sigma-Aldrich

Magna ChIRP U2snRNA Probe

The Magna ChIRP U2snRNA contains a 20-mer DNA oligonucleotide complementary to the sequence of Human RNA, U2 small nuclear 1.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

manufacturer/tradename

Magna ChIRP®

technique(s)

DNA sequencing: suitable
RNA extraction: suitable (nuclear function kit)
RNA purification: suitable (with magnetic beads)
activity assay: suitable (protein interaction)
nucleic acid detection: suitable (RNA)

NCBI accession no.

General description

Pervasive genomic transcription and the role of non-coding RNAs in this process is a rapidly evolving field that remains to be thoroughly explored in gene regulation.
Historically, chromatin immunoprecipitation (ChIP) has been used to interrogate association of proteins with genomic DNA sequences. Given that chromatin contains not only DNA and DNA-binding proteins, but also RNA-binding proteins and RNA, new methods have been developed to map the association of these RNA molecules to distinct regions of the genome. One such method, designated ChIRP (Chromatin Isolation by RNA Purification), allows recovery of specific segments of DNA and proteins using chromatin associated RNA as a target. This unique approach allows isolation of chromatin-associated RNAs (e.g. lncRNAs) within specific regions of genomic DNA sequences, as well as proteomic analysis of protein components of the lncRNA regulatory complexes. The Magna ChIRP® RNA Interactome Kits (cat. # 17-10494 and 17-10495) facilitate the use of this method and has been proven compatible with this probe set.
The Magna ChIRP U2snRNA contains a 20-mer DNA oligonucleotide complementary to the sequence of Human RNA, U2 small nuclear 1. The probe is biotin-TEG-labeled at the 3-prime end. The oligonucleotide is supplied ready for use in the ChIRP method.

Application

The Magna ChIRP U2snRNA contains a 20-mer DNA oligonucleotide complementary to the sequence of Human RNA, U2 small nuclear 1.

Quality

Verified by Electrospray Ionization Mass Spectrometry. Final yields are determined using UV absorbance at OD260. Concentrations are determined using UV absorbance at OD260 before pooling. Oligos will manufactured under ISO 9001 conditions.

Physical form

10 mM Tris-HCl, 1mM EDTA, pH 8.0

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

MAGNA CHIRP is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service