03-0195-00
SMC® Human IL-2 High Sensitivity Immunoassay Kit
Select a Size
$1,600.00
Select a Size
About This Item
$1,600.00
Recommended Products
grade
for protein analysis
feature
accuracy Minimum of 5 plasma samples spiked with average recovery of 83% with a range of 78-87%
accuracy Minimum of 5 serum samples spiked with average recovery of 91% with a range of 79-99%
sensitivity measuring range
0.09 pg/mL (Lower Limit of Quantification)
technique(s)
single molecule counting: suitable
General description
Application
- Improved options for sample prep and incubation
- Lyophilization of the standard provides the kit at one storage condition
- Increased reagent volumes to accommodate automation
- Validated to run on both TM=["SMCxPRO"] and Erenna
Components
Other Notes
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Use the power of high sensitivity immunology assays with SMC® technology to precisely characterize low-abundant cytokines like those from Th1/Th2 and Th17 cells.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service