Skip to Content
MilliporeSigma
All Photos(1)

Documents

03-0185-00

Millipore

SMC® Human Total Tau High Sensitivity Immunoassay Kit

Synonym(s):

Microtubile-associated protein tau immunoassay kit, Neurofibrillary tangle protein immunoassay kit, Tau Single Molecule Counting Kit, Tau ultra-sensitive immunoassay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.77

species reactivity

human

manufacturer/tradename

SMC®

assay range

sensitivity: 4.69 pg/mL
((lower limit of quantification))

inter-assay cv: ≤4%
intra-assay cv: ≤8%
(Intra-assay CV n=5; Inter-assay n=3)

General description

The human Tau family of microtubule-associated neuronal proteins has been rigorously studied in the context of normal and aberrant neurobiology. While typically involved in the stability of axonal microtubules, Tau proteins can also play a role in neurodegenerative diseases such as Alzheimer′s Disease (AD) and frontotemporal dementia (FTD) if they aberrantly misfold and accumulate in the brain. There is significant interest in studying the level of Tau proteins in both the central nervous system as well as in circulation, and the new SMC® Human Total-Tau High Sensitivity Immunoassay Kit enables researchers to take advantage of the Single Molecule Counting ultrasensitive immunoassay technology to perform accurately and sensitive measurements in human cerebrospinal fluid (CSF), plasma, and serum samples, toward making important discoveries about the role of Tau protein in neurodegenerative disease mechanisms.
The SMC® Human Total Tau High Sensitivity Immunoassay Kit contains all reagents required to perform the assay. This assay takes advantage of a standard immunoassay workflow configured in a standard plate format. Assay protocols are similar to existing sandwich ELISA methods with two key differences:
1) Elution buffer disrupts the sandwich, separating the labeled detection antibody for quantification.
2) The Erenna and TM="SMCxPRO"] immunoassay systems detect analytes using Single Molecule Counting SMC® technology.

Specificity

Specificity to the following species samples were tested with the follow results:
  • Non Human Primate – 50%
  • Mouse – 100%
  • Rat – 100%
  • Canine – 100%
  • Feline – 75%

Application

The sensitivity of the Single Molecule Counting SCM technology gives the SCM Human Total Tau High Sensitivity Immunoassay Kit unparalleled performance in the quantification of Human Total Tau in human serum, plasma, and CSF.

This new streamline kit has the following features:
  • Improved options for sample prep and incubation
  • Lyophilization protein standard for uniform shipping and storage condition
  • Increased reagent volumes to accommodate automation
  • Able to achieve assay specifications on both SMCxPRO® and Erenna.
The sensitivity of the Single Molecule Counting SMC® technology gives the SMC® Human Total Tau High Sensitivity Immunoassay Kit unparalleled performance in the quantification of Human Total Tau in human serum, plasma, and CSF.

Research Category:
Neuroscience
neurological

Research Sub-Category:
Alzheimer′s Disease
Aging, Nervous System
Neurodegenerative Diseases
Neurofilament & Neuron Metabolism
Neuronal & Glial Markers
Neuroregenerative Medicine
Neuroscience

Components

  • Assay Buffer
  • Total Tau Coated Beads
  • Standard Diluent
  • Total Tau Detection Antibody
  • Total Tau Standard
  • 10X Wash Buffer
  • Buffer D
  • Elution Buffer B

Specifications

Minimum of 3 serum samples spiked with average recovery of 70% with a range of 64-80%
Minimum of 4 plasma samples spiked with average recovery of 82% with a range of 76-88%
Minimum of 5 CSF samples spiked with average recovery of 91% with a range of 85-106%

Legal Information

SMC is a registered trademark of Singulex
SMCxPRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Neuroscience biomarker research expands with high-sensitivity immunoassays for low-abundant biomarkers.

Explore high-sensitivity SMC® neurology immunoassays that precisely measure low levels of neurology protein analytes, such as NF-L or Tau proteins.

Find immunoassays to explore neuroimmunology and neuroinflammation cytokines and neurodegenerative disease biomarkers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service