Select a Size
$2,170.00
Select a Size
About This Item
$2,170.00
Recommended Products
technique(s)
single molecule counting: suitable
General description
Application
- Five 96-well Assays Per Kit: Test on multiple washers or before and after plate washer maintenance<br />
- Abbreviated Workflow: Execute your assay in approximately 1 hour<br />
- Evaluates both the TM=["BioTek"] 405 TS and Tecan HydroFlex microplate washers<br />
- Data analyzable on both the TM=["SMCxPRO"] and Erenna platforms<br />
- Confirm optimal plate washer performance before evaluating your precious samples
Components
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B - Skin Sens. 1
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Learn how to optimize your plate washer performance with the SMC® Plate Washer Evaluation Kit and assay precious samples with confidence.
Use the power of high sensitivity immunology assays with SMC® technology to precisely characterize low-abundant cytokines like those from Th1/Th2 and Th17 cells.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service