Skip to Content
MilliporeSigma
All Photos(1)

Documents

03-0128-00

Millipore

SMC® Plate-Based Immunoassay Development Kit

Synonym(s):

SMC® Assay Development Kit, Single Molecule Counting Assay Development Kit, Ultra-sensitive Assay Development Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

manufacturer/tradename

SMC®

Quality Level

technique(s)

ELISA: suitable
single molecule counting: suitable

General description

Take advantage of Single Molecule Counting (SMC®) technology while developing your own application-specific immunoassays in plate format. This kit provides everything you need to build your own high performing SMC® immunoassay. It includes buffers, reagents and optimized protocol for plate assay development.

Application

Create application-specific immunoassays in plate format and move beyond the traditional ELISA with SMC® Technology.
For use with the SMC® immunoassay system.

Components

Assay Buffer
Blocking Buffer
Plate Standard Diluent
10X Wash Buffer
Elution Buffer B
Buffer C
10X PBS

Legal Information

SMC is a registered trademark of Singulex

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service