Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01-170

Sigma-Aldrich

NGF 7S Protein, mouse

The mouse NGF 7S Protein( or nerve growth factor 7S) is available in a 100 µg format.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

biological source

mouse

manufacturer/tradename

Upstate®

technique(s)

cell culture | mammalian: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

mouse ... NGF(18049)

General description

Product Source: Male mouse submaxillary glands

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The nerve growth factor thirty-five years later.
Levi-Montalcini, R
In Vitro Cellular & Developmental Biology, 23, 227-238 (1987)
The biological activity of cross-linked beta nerve growth factor protein.
R W Stach et al.
The Journal of biological chemistry, 249(20), 6668-6674 (1974-10-25)
Nerve growth factor acts in brain.
Marx, J L
Science (New York, N.Y.), 232, 1341-1342 (1986)
Chuck C-K Chao et al.
Journal of neuroscience research, 74(2), 248-254 (2003-09-30)
Gas7, a growth arrest-specific gene originally isolated from serum-starved mouse fibroblast cells, is expressed in vivo predominantly in the brain and is required for neurite formation in cultured mouse cerebellar neurons (Ju et al. [1998] Proc. Natl. Acad. Sci. USA
M A Smith et al.
The Journal of biological chemistry, 272(25), 15675-15681 (1997-06-20)
The extracellular matrix protein agrin plays an important role in the formation and maintenance of the neuromuscular junction. However, regulation of agrin gene expression and pre-mRNA splicing, important in determining the biological actions of agrin, is not well understood. To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service