Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAT047030453

Matrix Scientific

4-Amino-N,N-diethyl-benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
228.32
MDL number:

mp

105-106

Iupac Name

4-amino-N,N-diethylbenzenesulfonamide

Inchi Code

1S/C10H16N2O2S/c1-3-12(4-2)15(13,14)10-7-5-9(11)6-8-10/h5-8H,3-4,11H2,1-2H3

InChI key

LTFVELCIFWEGGA-UHFFFAOYSA-N

Country of Origin

US

Product Link

https://www.matrixscientific.com/011459.html

Melting Point

105-106

Pictograms

MSDS

https://www.matrixscientific.com/011459.html


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service