Skip to Content
MilliporeSigma
All Photos(1)

Documents

KEY947335527

Key Organics / BIONET

3-(4-methylpiperazin-1-yl)propan-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
158.24
MDL number:

Iupac Name

3-(4-methyl-1-piperazinyl)-1-propanol

Inchi Code

1S/C8H18N2O/c1-9-4-6-10(7-5-9)3-2-8-11/h11H,2-8H2,1H3

InChI key

JKRSQNBRNIYETC-UHFFFAOYSA-N

storage temp.

Store under inert gas

Purity

95%

Country of Origin

GB

Physical Form

Liquid

Product Link

https://www.keyorganics.net/sku/PS-3230

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary statements

MSDS

https://www.keyorganics.net/sds/PS-3230


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service