Skip to Content
MilliporeSigma
All Photos(1)

Documents

KEY458251329

Key Organics / BIONET

3-bromo-4-(morpholin-4-yl)aniline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
257.13
MDL number:

Iupac Name

3-bromo-4-(4-morpholinyl)aniline

Inchi Code

1S/C10H13BrN2O/c11-9-7-8(12)1-2-10(9)13-3-5-14-6-4-13/h1-2,7H,3-6,12H2

InChI key

IWWZCFFYGZRVHR-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/11P-075

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/11P-075


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service