Skip to Content
MilliporeSigma
All Photos(1)

Documents

JWPH3249E79F

J & W Pharmlab LLC

2-Phenyl-octahydro-pyrrolo[3,4-c]pyrrole dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
261.19
MDL number:

Iupac Name

2-phenyloctahydropyrrolo[3,4-c]pyrrole dihydrochloride

Inchi Code

1S/C12H16N2.2ClH/c1-2-4-12(5-3-1)14-8-10-6-13-7-11(10)9-14;;/h1-5,10-11,13H,6-9H2;2*1H

InChI key

ARVBADVNKVWNEY-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Physical Form

Light Grey Solid

Product Link

https://www.jwpharmlab.com/home/product/75R0165S

MSDS

https://www.jwpharmlab.com/home/product/75R0165S


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service