Skip to Content
MilliporeSigma
All Photos(1)

Documents

JWPH3249CA00

J & W Pharmlab LLC

2-tert-Butoxycarbonylamino-3-oxo-3-phenyl-propionic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
307.35
MDL number:

Iupac Name

ethyl 2-[(tert-butoxycarbonyl)amino]-3-oxo-3-phenylpropanoate

Inchi Code

1S/C16H21NO5/c1-5-21-14(19)12(17-15(20)22-16(2,3)4)13(18)11-9-7-6-8-10-11/h6-10,12H,5H2,1-4H3,(H,17,20)

InChI key

DVPJXQCIYYLQGB-UHFFFAOYSA-N

Purity

96%

Country of Origin

US

Physical Form

Off White Solid

Product Link

https://www.jwpharmlab.com/home/product/20R0593

MSDS

https://www.jwpharmlab.com/home/product/20R0593


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service