Iupac Name
3-oxa-7-azabicyclo[4.1.0]heptane
Inchi Code
1S/C5H9NO/c1-2-7-3-5-4(1)6-5/h4-6H,1-3H2
InChI key
DIUNZTQKLKGEBB-UHFFFAOYSA-N
storage temp.
-10
Purity
91
Country of Origin
UA
Shipping Temperature
Ice Pack
Physical Form
liquid
Product Link
https://www.enaminestore.com/catalog/EN300-300958
Signal Word
Danger
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-300958
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service