Skip to Content
MilliporeSigma
All Photos(1)

Documents

ENA238712106

Enamine

5-methyl-2,3,4,5-tetrahydro-1,4-benzoxazepine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
163.22
MDL number:

Iupac Name

5-methyl-2,3,4,5-tetrahydro-1,4-benzoxazepine

Inchi Code

1S/C10H13NO/c1-8-9-4-2-3-5-10(9)12-7-6-11-8/h2-5,8,11H,6-7H2,1H3

InChI key

BDMKKDLAKZPRIO-UHFFFAOYSA-N

storage temp.

4

Purity

80

Country of Origin

UA

Shipping Temperature

Ice Pack

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-50113

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-50113


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service