CIAH987ED208
ChemScene LLC (Preferred Partner)
2-Amino-5-fluoropyridin-3-ol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
bp
370.9+-42.0 C at 760 mmHg
Iupac Name
2-amino-5-fluoro-3-pyridinol
Inchi Code
1S/C5H5FN2O/c6-3-1-4(9)5(7)8-2-3/h1-2,9H,(H2,7,8)
InChI key
QEDZENLHGZUFBK-UHFFFAOYSA-N
storage temp.
4C, protect from light
Purity
98%
Country of Origin
CN
Physical Form
Solid
Product Link
https://www.chemscene.com/1003711-04-3.html
Boiling point
370.9+-42.0 C at 760 mmHg
Signal Word
Danger
Hazard Statements
Precautionary statements
MSDS
https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0130932.pdf
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service