Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V-076

Supelco

Vilazodone hydrochloride solution

1 mg/mL (Methanol:DMSO (90:10) (v/v)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$241.00

$241.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$241.00

About This Item

Empirical Formula (Hill Notation):
C26H27N5O2 · HCl
CAS Number:
Molecular Weight:
477.99
MDL number:
UNSPSC Code:
41116107

$241.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL (Methanol:DMSO (90:10) (v/v))

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

N#CC1=CC=C(NC=C2CCCCN3CCN(C4=CC(C=C(C(N)=O)O5)=C5C=C4)CC3)C2=C1.Cl

InChI

1S/C26H27N5O2.ClH/c27-16-18-4-6-23-22(13-18)19(17-29-23)3-1-2-8-30-9-11-31(12-10-30)21-5-7-24-20(14-21)15-25(33-24)26(28)32;/h4-7,13-15,17,29H,1-3,8-12H2,(H2,28,32);1H

InChI key

RPZBRGFNBNQSOP-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Vilazodone, marketed as the antidepressant drug Viibryd®, is a serotonin reuptake inhibitor for treatment of major depressive disorder. Suitable uses for this Certified Spiking Solution® include as starting material for calibrators and controls in antidepressant testing methods by LC/MS or
GC/MS for clinical toxicology, urine drug testing, or forensic analysis applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
VIIBRYD is a registered trademark of Forest Laboratories, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service