Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T-170

Supelco

(-)-11-nor-9-Carboxy-delta8-THC solution

1 mg/mL (Degassed Methanol), certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

11-Nor-Δ8-tetrahydrocannabinol-9-carboxylic acid solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$259.00

$259.00


Available to ship onApril 23, 2025Details


Request a Bulk Order

Select a Size

Change View
1 ML
$259.00

About This Item

Empirical Formula (Hill Notation):
C21H28O4
CAS Number:
Molecular Weight:
344.44
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$259.00


Available to ship onApril 23, 2025Details


Request a Bulk Order

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL (Degassed Methanol)

format

single component solution

storage temp.

-10 to -25°C

SMILES string

CCCCCc1cc(O)c2[C@@H]3CC(=CC[C@H]3C(C)(C)Oc2c1)C(O)=O

InChI

1S/C21H28O4/c1-4-5-6-7-13-10-17(22)19-15-12-14(20(23)24)8-9-16(15)21(2,3)25-18(19)11-13/h8,10-11,15-16,22H,4-7,9,12H2,1-3H3,(H,23,24)/t15-,16-/m1/s1

InChI key

OGXXAQFMAFTSRU-HZPDHXFCSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service