Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T-114

Supelco

(-)-delta9-THC-O-beta-D-glucuronide sodium salt solution

100 μg/mL (Degassed Methanol, (as free carboxylate)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$370.00

$370.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$370.00

About This Item

Empirical Formula (Hill Notation):
C27H37NaO8
Molecular Weight:
512.57
UNSPSC Code:
41116107
NACRES:
NA.24

$370.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

description

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (Degassed Methanol, (as free carboxylate))

format

single component solution

storage temp.

-10 to -25°C

General description

(-)-Delta9-THC-beta-D-glucuronide is a major urinary metabolite of Delta9-THC. This certified Snap-N-Spike® reference solution is suitable for numerous GC/MS or LC/MS applications from pharmaceutical testing and research to clinical toxicology or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service